Assay Details
| Gene Symbol: AT1G12050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101077.6 | NP_172669.2 | 12 - 13 | 1242 | 77 | |
| GenBank mRNA | AY094010.1 | - | 12 - 13 | 1079 | 77 | |
More Information
Set Membership: |
|

AT1G12050
Genomic Map