Assay Details
| Gene Symbol: CPN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332142.1 | NP_001322728.1 | 1 - 2 | 389 | 74 | |
| NM_101367.3 | NP_563961.1 | 1 - 2 | 389 | 74 | ||
| GenBank mRNA | AY063928.1 | - | 1 - 2 | 141 | 74 | |
| AY086708.1 | - | 1 - 2 | 195 | 74 | ||
| AY091252.1 | - | 1 - 2 | 104 | 74 | ||
| D88314.2 | - | 1 - 2 | 117 | 74 | ||
| L02843.1 | - | 1 - 2 | 117 | 74 | ||
More Information
Set Membership: |
|

CPN10
Genomic Map