Assay Details
| Gene Symbol: CPN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101367.3 | NP_563961.1 | 2 - 3 | 524 | 74 | |
| GenBank mRNA | AY063928.1 | - | 2 - 3 | 276 | 74 | |
| AY086708.1 | - | 2 - 3 | 330 | 74 | ||
| AY091252.1 | - | 2 - 3 | 239 | 74 | ||
| D88314.2 | - | 2 - 3 | 252 | 74 | ||
| L02843.1 | - | 2 - 3 | 252 | 74 | ||
More Information
Set Membership: |
|

CPN10
Genomic Map