Assay Details
| Gene Symbol: TIFY10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106153.4 | NP_565096.1 | 4 - 5 | 847 | 76 | |
| GenBank mRNA | AF344319.1 | - | 4 - 5 | 659 | 76 | |
| AF386959.1 | - | 4 - 5 | 795 | 76 | ||
| BT006301.1 | - | 4 - 5 | 659 | 76 | ||
More Information
Set Membership: |
|

TIFY10B
Genomic Map