Assay Details
| Gene Symbol: ASHH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334740.1 | NP_001321484.1 | 3 - 4 | 247 | 112 | |
| NM_001334741.1 | NP_001321485.1 | 3 - 4 | 212 | 112 | ||
| NM_106321.4 | NP_177797.2 | 3 - 4 | 230 | 112 | ||
| NM_202429.2 | NP_974158.1 | 2 - 3 | 359 | 112 | ||
| GenBank mRNA | AB493538.1 | - | 2 - 3 | 90 | 112 | |
| BT001913.1 | - | 2 - 3 | 90 | 112 | ||
More Information
Set Membership: |
|

ASHH1
Genomic Map