Assay Details
| Gene Symbol: AT1G77750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106424.4 | NP_177898.1 | 2 - 3 | 530 | 62 | |
| GenBank mRNA | BT010186.1 | - | 2 - 3 | 293 | 62 | |
More Information
Set Membership: |
|

AT1G77750
Genomic Map