Assay Details
| Gene Symbol: AGL66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106447.4 | NP_177921.2 | 1 - 2 | 302 | 92 | |
| GenBank mRNA | DQ446441.1 | - | 1 - 2 | 183 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL66| Gene Symbol: AGL66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106447.4 | NP_177921.2 | 1 - 2 | 302 | 92 | |
| GenBank mRNA | DQ446441.1 | - | 1 - 2 | 183 | 92 | |
Set Membership: |
|