Assay Details
| Gene Symbol: AGL66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106447.4 | NP_177921.2 | 8 - 9 | 661 | 101 | |
| GenBank mRNA | DQ446441.1 | - | 8 - 9 | 542 | 101 | |
More Information
Set Membership: |
|

AGL66
Genomic Map