Assay Details
| Gene Symbol: TPR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334819.1 | NP_001322855.1 | 2 - 3 | 1014 | 113 | |
| NM_106462.3 | NP_177936.1 | 2 - 3 | 1014 | 113 | ||
More Information
Set Membership: |
|

TPR12
Genomic Map