Assay Details
| Gene Symbol: GGH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198500.1 | NP_001185429.1 | 2 - 3 | 586 | 113 | |
| NM_106515.4 | NP_565186.2 | 2 - 3 | 561 | 113 | ||
| GenBank mRNA | AY070047.1 | - | 2 - 3 | 477 | 113 | |
| AY096428.1 | - | 2 - 3 | 409 | 113 | ||
More Information
Set Membership: |
|

GGH2
Genomic Map