Assay Details
| Gene Symbol: FTSH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334887.1 | NP_001321597.1 | 13 - 14 | 2735 | 72 | |
| NM_001334888.1 | NP_001321596.1 | 13 - 14 | 2735 | 72 | ||
| NM_106604.4 | NP_565212.1 | 13 - 14 | 2600 | 72 | ||
| GenBank mRNA | AK317610.1 | - | 13 - 14 | 2566 | 72 | |
| AX765796.1 | - | 13 - 14 | 2383 | 72 | ||
More Information
Set Membership: |
|

FTSH12
Genomic Map