Assay Details
| Gene Symbol: FTSH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334887.1 | NP_001321597.1 | 7 - 8 | 1933 | 111 | |
| NM_001334888.1 | NP_001321596.1 | 7 - 8 | 1933 | 111 | ||
| NM_106604.4 | NP_565212.1 | 7 - 8 | 1798 | 111 | ||
| GenBank mRNA | AK317610.1 | - | 7 - 8 | 1764 | 111 | |
| AX765796.1 | - | 7 - 8 | 1581 | 111 | ||
More Information
Set Membership: |
|

FTSH12
Genomic Map