Assay Details
| Gene Symbol: SYP52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036224.2 | NP_001031301.1 | 6 - 7 | 748 | 92 | |
| NM_001334895.1 | NP_001321707.1 | 5 - 6 | 773 | 92 | ||
| NM_106607.5 | NP_565213.1 | 5 - 6 | 853 | 92 | ||
| GenBank mRNA | AF355756.1 | - | 5 - 6 | 742 | 92 | |
| BT003941.1 | - | 5 - 6 | 724 | 92 | ||
| BT006106.1 | - | 4 - 5 | 566 | 92 | ||
More Information
Set Membership: |
|

SYP52
Genomic Map