Assay Details
| Gene Symbol: NUDT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334912.1 | NP_001320544.1 | 20 - 21 | 2261 | 66 | |
| NM_106618.5 | NP_565218.1 | 19 - 20 | 2433 | 66 | ||
| GenBank mRNA | AY120774.1 | - | 19 - 20 | 2149 | 66 | |
| BT000142.1 | - | 19 - 20 | 2119 | 66 | ||
More Information
Set Membership: |
|

NUDT3
Genomic Map