Assay Details
| Gene Symbol: NADP-ME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106624.5 | NP_178093.1 | 12 - 13 | 1797 | 70 | |
| GenBank mRNA | AY060584.1 | - | 12 - 13 | 1475 | 70 | |
| AY142064.1 | - | 12 - 13 | 1286 | 70 | ||
More Information
Set Membership: |
|

NADP-ME4
Genomic Map