Assay Details
| Gene Symbol: NADP-ME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106624.5 | NP_178093.1 | 16 - 17 | 2145 | 72 | |
| GenBank mRNA | AY060584.1 | - | 16 - 17 | 1823 | 72 | |
| AY142064.1 | - | 16 - 17 | 1634 | 72 | ||
More Information
Set Membership: |
|

NADP-ME4
Genomic Map