Assay Details
| Gene Symbol: NADP-ME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106624.5 | NP_178093.1 | 2 - 3 | 945 | 119 | |
| GenBank mRNA | AY060584.1 | - | 2 - 3 | 623 | 119 | |
| AY142064.1 | - | 2 - 3 | 434 | 119 | ||
More Information
Set Membership: |
|

NADP-ME4
Genomic Map