Assay Details
| Gene Symbol: AT1G80850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106733.3 | NP_178200.1 | 2 - 3 | 818 | 119 | |
| GenBank mRNA | BT033081.1 | - | 2 - 3 | 488 | 119 | |
More Information
Set Membership: |
|

AT1G80850
Genomic Map