Assay Details
| Gene Symbol: AT1G80910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106739.4 | NP_178206.1 | 4 - 5 | 791 | 77 | |
| GenBank mRNA | AK318688.1 | - | 4 - 5 | 679 | 77 | |
More Information
Set Membership: |
|

AT1G80910
Genomic Map