Assay Details
| Gene Symbol: UBC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202912.1 | NP_001189841.1 | 3 - 4 | 527 | 123 | |
| NM_111703.4 | NP_566331.1 | 3 - 4 | 431 | 123 | ||
| GenBank mRNA | AK228254.1 | - | 3 - 4 | 301 | 123 | |
| AY063869.1 | - | 3 - 4 | 294 | 123 | ||
| AY091223.1 | - | 2 - 3 | 198 | 123 | ||
| DQ027025.1 | - | 2 - 3 | 198 | 123 | ||
More Information
Set Membership: |
|

UBC11
Genomic Map