Assay Details
| Gene Symbol: APX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035587.3 | NP_001030664.1 | 4 - 5 | 424 | 75 | |
| NM_111798.4 | NP_187575.2 | 3 - 4 | 507 | 75 | ||
| GenBank mRNA | AK176821.1 | - | 4 - 5 | 424 | 75 | |
| AK176908.1 | - | 3 - 4 | 507 | 75 | ||
More Information
Set Membership: |
|

APX2
Genomic Map