Assay Details
| Gene Symbol: APX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035587.3 | NP_001030664.1 | 8 - 9 | 738 | 93 | |
| NM_111798.4 | NP_187575.2 | 7 - 8 | 821 | 93 | ||
| GenBank mRNA | AK176821.1 | - | 8 - 9 | 738 | 93 | |
| AK176908.1 | - | 7 - 8 | 821 | 93 | ||
More Information
Set Membership: |
|

APX2
Genomic Map