Assay Details
| Gene Symbol: ADK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111817.4 | NP_187593.1 | 5 - 6 | 666 | 70 | |
| NM_202540.1 | NP_974269.1 | 4 - 5 | 643 | 70 | ||
| GenBank mRNA | AF180894.1 | - | 5 - 6 | 373 | 70 | |
| AF375451.1 | - | 5 - 6 | 393 | 70 | ||
| BT033101.1 | - | 3 - 4 | 198 | 70 | ||
More Information
Set Membership: |
|

ADK1
Genomic Map