Assay Details
| Gene Symbol: OMR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111840.3 | NP_187616.1 | 1 - 2 | 714 | 132 | |
| GenBank mRNA | AF096281.1 | - | 1 - 2 | 407 | 132 | |
| AY065037.1 | - | 1 - 2 | 426 | 132 | ||
More Information
Set Membership: |
|

OMR1
Genomic Map