Assay Details
| Gene Symbol: OMR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111840.3 | NP_187616.1 | 3 - 4 | 1169 | 63 | |
| GenBank mRNA | AF096281.1 | - | 3 - 4 | 862 | 63 | |
| AY065037.1 | - | 3 - 4 | 881 | 63 | ||
More Information
Set Membership: |
|

OMR1
Genomic Map