Assay Details
| Gene Symbol: OMR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111840.3 | NP_187616.1 | 4 - 5 | 1365 | 78 | |
| GenBank mRNA | AF096281.1 | - | 4 - 5 | 1058 | 78 | |
| AY065037.1 | - | 4 - 5 | 1077 | 78 | ||
More Information
Set Membership: |
|

OMR1
Genomic Map