Assay Details
| Gene Symbol: RPL12-C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113701.3 | NP_189423.1 | 1 | 687 | 131 | |
| GenBank mRNA | AY037241.1 | - | 1 | 632 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL12-C| Gene Symbol: RPL12-C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113701.3 | NP_189423.1 | 1 | 687 | 131 | |
| GenBank mRNA | AY037241.1 | - | 1 | 632 | 131 | |
Set Membership: |
|