Assay Details
| Gene Symbol: AT3G47840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114654.2 | NP_190368.1 | 1 | 2130 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G47840| Gene Symbol: AT3G47840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114654.2 | NP_190368.1 | 1 | 2130 | 101 | |
Set Membership: |
|