Assay Details
| Gene Symbol: MNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114947.3 | NP_566939.1 | 1 - 2 | 679 | 78 | |
| GenBank mRNA | AB493644.1 | - | 1 - 2 | 514 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MNP| Gene Symbol: MNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114947.3 | NP_566939.1 | 1 - 2 | 679 | 78 | |
| GenBank mRNA | AB493644.1 | - | 1 - 2 | 514 | 78 | |
Set Membership: |
|