Assay Details
| Gene Symbol: VPS35C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114990.5 | NP_190699.3 | 3 - 4 | 436 | 91 | |
| GenBank mRNA | AB368490.1 | - | 3 - 4 | 303 | 91 | |
More Information
Set Membership: |
|

VPS35C
Genomic Map