Assay Details
| Gene Symbol: EMB1691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084896.2 | NP_001078365.1 | 1 - 2 | 2285 | 98 | |
| NM_117066.4 | NP_567348.2 | 1 - 2 | 2306 | 98 | ||
| GenBank mRNA | AY046016.1 | - | 1 - 2 | 2280 | 98 | |
| BT000992.1 | - | 1 - 2 | 2022 | 98 | ||
More Information
Set Membership: |
|

EMB1691
Genomic Map