Assay Details
| Gene Symbol: EMB1691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084896.2 | NP_001078365.1 | 4 - 5 | 2602 | 89 | |
| GenBank mRNA | AY046016.1 | - | 4 - 5 | 2597 | 89 | |
More Information
Set Membership: |
|

EMB1691
Genomic Map