Assay Details
| Gene Symbol: SDG16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118929.5 | NP_194520.3 | 22 - 23 | 3766 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDG16| Gene Symbol: SDG16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118929.5 | NP_194520.3 | 22 - 23 | 3766 | 71 | |
Set Membership: |
|