Assay Details
| Gene Symbol: AT4G28080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118947.8 | NP_194537.7 | 18 - 19 | 3248 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G28080| Gene Symbol: AT4G28080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118947.8 | NP_194537.7 | 18 - 19 | 3248 | 78 | |
Set Membership: |
|