Assay Details
| Gene Symbol: RBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161253.1 | NP_001154725.1 | 2 - 3 | 280 | 64 | |
| NM_001161254.1 | NP_001154726.1 | 2 - 3 | 334 | 64 | ||
| NM_122064.3 | NP_568396.1 | 2 - 3 | 307 | 64 | ||
| GenBank mRNA | AK118181.1 | - | 2 - 3 | 271 | 64 | |
| AY052401.1 | - | 2 - 3 | 190 | 64 | ||
| AY072430.1 | - | 2 - 3 | 277 | 64 | ||
| AY086913.1 | - | 2 - 3 | 280 | 64 | ||
| AY114719.1 | - | 2 - 3 | 190 | 64 | ||
More Information
Set Membership: |
|

RBX1
Genomic Map