Assay Details
| Gene Symbol: RBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161253.1 | NP_001154725.1 | 5 - 6 | 485 | 92 | |
| NM_001161254.1 | NP_001154726.1 | 4 - 5 | 467 | 92 | ||
| NM_122064.3 | NP_568396.1 | 4 - 5 | 440 | 92 | ||
| GenBank mRNA | AK118181.1 | - | 4 - 5 | 404 | 92 | |
| AY072430.1 | - | 4 - 5 | 410 | 92 | ||
| AY086913.1 | - | 4 - 5 | 413 | 92 | ||
| AY114719.1 | - | 4 - 5 | 323 | 92 | ||
More Information
Set Membership: |
|

RBX1
Genomic Map