Assay Details
| Gene Symbol: BGAL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343683.1 | NP_001330417.1 | 1 - 2 | 372 | 68 | |
| NM_122078.5 | NP_568399.4 | 4 - 5 | 966 | 68 | ||
More Information
Set Membership: |
|

BGAL7
Genomic Map