Assay Details
| Gene Symbol: BGAL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343683.1 | NP_001330417.1 | 5 - 6 | 1137 | 86 | |
| NM_122078.5 | NP_568399.4 | 8 - 9 | 1731 | 86 | ||
More Information
Set Membership: |
|

BGAL7
Genomic Map