Assay Details
| Gene Symbol: JAZ12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122098.5 | NP_197590.1 | 3 - 4 | 587 | 64 | |
| GenBank mRNA | AF360184.1 | - | 3 - 4 | 502 | 64 | |
| AY051013.1 | - | 3 - 4 | 408 | 64 | ||
More Information
Set Membership: |
|

JAZ12
Genomic Map