Assay Details
| Gene Symbol: RAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122181.3 | NP_197667.1 | 12 - 13 | 2248 | 126 | |
| GenBank mRNA | AK228695.1 | - | 12 - 13 | 2222 | 126 | |
More Information
Set Membership: |
|

RAD5
Genomic Map