Assay Details
| Gene Symbol: RAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122181.3 | NP_197667.1 | 1 - 2 | 771 | 73 | |
| GenBank mRNA | AK228695.1 | - | 1 - 2 | 745 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAD5| Gene Symbol: RAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122181.3 | NP_197667.1 | 1 - 2 | 771 | 73 | |
| GenBank mRNA | AK228695.1 | - | 1 - 2 | 745 | 73 | |
Set Membership: |
|