Assay Details
| Gene Symbol: RAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122181.3 | NP_197667.1 | 6 - 7 | 1397 | 101 | |
| GenBank mRNA | AK228695.1 | - | 6 - 7 | 1371 | 101 | |
More Information
Set Membership: |
|

RAD5
Genomic Map