Assay Details
| Gene Symbol: MGT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343773.1 | NP_001332159.1 | 5 - 6 | 795 | 78 | |
| NM_122188.3 | NP_568424.1 | 5 - 6 | 795 | 78 | ||
| GenBank mRNA | BT029184.1 | - | 5 - 6 | 716 | 78 | |
More Information
Set Membership: |
|

MGT10
Genomic Map