Assay Details
| Gene Symbol: TAF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343910.1 | NP_001330252.1 | 16 - 17 | 1837 | 66 | |
| NM_122424.4 | NP_197897.3 | 16 - 17 | 1854 | 66 | ||
| GenBank mRNA | AY463620.1 | - | 16 - 17 | 1786 | 66 | |
More Information
Set Membership: |
|

TAF5
Genomic Map