Assay Details
| Gene Symbol: Cpn60beta4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198168.2 | NP_001185097.1 | 8 - 9 | 1225 | 71 | |
| NM_001332699.1 | NP_001319083.1 | 9 - 10 | 1365 | 71 | ||
| NM_001332700.1 | NP_001322215.1 | 9 - 10 | 1361 | 71 | ||
| NM_102387.3 | NP_173947.1 | 9 - 10 | 1408 | 71 | ||
More Information
Set Membership: |
|

Cpn60beta4
Genomic Map