Assay Details
| Gene Symbol: CAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102395.3 | NP_564243.1 | 4 - 5 | 575 | 90 | |
| GenBank mRNA | AB493477.1 | - | 4 - 5 | 441 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAL| Gene Symbol: CAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102395.3 | NP_564243.1 | 4 - 5 | 575 | 90 | |
| GenBank mRNA | AB493477.1 | - | 4 - 5 | 441 | 90 | |
Set Membership: |
|