Assay Details
| Gene Symbol: BGLU40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102418.4 | NP_173978.1 | 8 - 9 | 1411 | 66 | |
| GenBank mRNA | AY045927.1 | - | 8 - 9 | 1245 | 66 | |
| AY142610.1 | - | 8 - 9 | 1110 | 66 | ||
More Information
Set Membership: |
|

BGLU40
Genomic Map