Assay Details
| Gene Symbol: AT1G26640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102426.6 | NP_173986.2 | 4 - 5 | 570 | 107 | |
| GenBank mRNA | AY150412.1 | - | 4 - 5 | 434 | 107 | |
More Information
Set Membership: |
|

AT1G26640
Genomic Map