Assay Details
| Gene Symbol: AT1G27520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102516.4 | NP_564288.1 | 3 - 4 | 424 | 101 | |
| GenBank mRNA | AY056382.1 | - | 3 - 4 | 326 | 101 | |
More Information
Set Membership: |
|

AT1G27520
Genomic Map