Assay Details
| Gene Symbol: AT1G28510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102619.4 | NP_564311.1 | 3 - 4 | 571 | 72 | |
| GenBank mRNA | AF412051.1 | - | 3 - 4 | 492 | 72 | |
| AY072613.1 | - | 3 - 4 | 352 | 72 | ||
| AY085695.1 | - | 3 - 4 | 499 | 72 | ||
More Information
Set Membership: |
|

AT1G28510
Genomic Map